missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IGF2BP2 Recombinant Human Protein, N-His.GST Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human IGF2BP2 isoform 1 (Q9Y6M1-2) (Met 1-Lys 599) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus

$147.00 - $590.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IMP-2, IMP2, VICKZ2, p62
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11116H20B5 View Documents Life Technologies
LS11116H20B5
5μg Each for $147.00
 
11116H20B50 View Documents Life Technologies
LS11116H20B50
50μg Each for $590.00
 
Description

Description

Insulin-like Growth Factor 2 mRNA Binding Protein 2 (IGF2BP2), recombinant human protein is supplied as a lyophilized powder.
  • Activity: This recombinant protein has not been tested
  • Formulation: 50mM Tris, 500mM NaCl , 0.5mM PMSF, 5mM GSH, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
IMP-2, IMP2, VICKZ2, p62
PAGE
Lyophilized
10644

For Research Use Only. Not for use in diagnostic procedures.