Please login to your online account to display your discounted pricing

Sino Biological™ IGF2BP2 Recombinant Human Protein, N-His.GST Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human IGF2BP2 isoform 1 (Q9Y6M1-2) (Met 1-Lys 599) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus

Manufacturer: life technologies  LS11116H20B5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11116H20B5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 94kDa
Purification Method PAGE
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Quantity 5μg
Gene ID (Entrez) 10644
Protein Family Growth Factors & Receptors
Protein Tag GST Tag/His Tag
Validated Application Assay Standard
Expression System Insect
Gene Alias IMP-2, IMP2, VICKZ2, p62
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Endotoxin Concentration <1 EU/μg
Common Name Insulin-like Growth Factor 2 mRNA Binding Protein 2
Structural Form Full Length/Recombinant
Gene Symbol IGF2BP2
Purity ≥90% by SDS-PAGE
Termination N-Terminal
Format Lyophilized

Insulin-like Growth Factor 2 mRNA Binding Protein 2 (IGF2BP2), recombinant human protein is supplied as a lyophilized powder.

  • Activity: This recombinant protein has not been tested
  • Formulation: 50mM Tris, 500mM NaCl , 0.5mM PMSF, 5mM GSH, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.