missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IGF2 Recombinant Human Protein, His.NusA Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IGF2 (P01344-1) (Ala 25-Glu 91) fused to the polyhistidine-NusA tag at the N-terminus

$180.00 - $180.00

Specifications

Purity ≥85% by SDS-PAGE
Format Lyophilized
Gene Alias PP1446, C11orf43, FLJ22066, FLJ44734, IGF-II, PP9974
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
13032H24E25 View Documents Life Technologies
LS13032H24E25
5 x 5μg Each for $180.00
 
Description

Description

Insulin-like growth factor 2 (IGF2), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IGF2 at 10μg/mL (100μL/well) can bind biotinylated human His-IGFBP3; the EC50 of binding to biotinylated human His-IGFBP3 is 18ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
PP1446, C11orf43, FLJ22066, FLJ44734, IGF-II, PP9974

For Research Use Only. Not for use in diagnostic procedures.