missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IGF1 Recombinant Human Protein, His.NusA Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IGF1 isoform 1 (P05019-1) (Gly 49-Ala 118) fused to the polyhistidine-NusA tag at the N-terminus

$80.50 - $147.00

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias IGF-I, IGF1A, IGFI
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10598H24E25 View Documents Life Technologies
LS10598H24E25
5 x 5μg Each for $112.00
 
10598H24E5 View Documents Life Technologies
LS10598H24E5
5μg Each for $80.50
 
10598H24E50 View Documents Life Technologies
LS10598H24E50
50μg Each for $147.00
 
Description

Description

Insulin-like growth factor 1 (IGF1), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IGF1 at 10μg/mL (100μL/well) can bind biotinylated human IGFBP3-His, the EC50 of biotinylated human IGFBP3-His is 24ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
IGF-I, IGF1A, IGFI

For Research Use Only. Not for use in diagnostic procedures.