Please login to your online account to display your discounted pricing

Sino Biological™ IGF1 Recombinant Human Protein, His.NusA Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form of human IGF1 isoform 1 (P05019-1) (Gly 49-Ala 118) fused to the polyhistidine-NusA tag at the N-terminus

Manufacturer: life technologies  LS10598H24E50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10598H24E50

  • / Each
 

Description & Specifications

Specifications

Quantity 50μg
Validated Application Assay Standard
Protein Family Growth Factors & Receptors
Protein Subtype IGFs (Insulin-Like Growth Factors)
Molecular Weight 64.8kDa
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Structural Form Full Length/Recombinant
Expression System E. coli
Common Name Insulin-like growth factor 1 (somatomedin C)
Gene Alias IGF-I, IGF1A, IGFI
Gene Symbol IGF1
Protein Tag His Tag/NusA Tag
Purity ≥90% by SDS-PAGE
Format Lyophilized

Insulin-like growth factor 1 (IGF1), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IGF1 at 10μg/mL (100μL/well) can bind biotinylated human IGFBP3-His, the EC50 of biotinylated human IGFBP3-His is 24ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.