missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IFNL3 (IL28B) Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human IFNL3 (NP_742151.2) (Met 1-Val 196) fused to a polyhistidine tag at the C-terminus

$618.00 - $618.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IFNL3, IL-28B, IL28C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11890H08H50 View Documents Life Technologies
LS11890H08H50
50μg Each for $618.00
 
Description

Description

Interferon Lambda 3 (IFNL3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: 1. Measured in anti-viral assays using WISH human amnion cells infected with vesicular stomatitis virus (VSV), the EC50 for this effect is typically 4-20ng/mL; 2. Measured by its binding ability in a functional ELISA; immobilized human IL28B at 20μg/mL (100μL/well) can bind human IL10RB with a linear ranger of 3.125-25μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, 500mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
IFNL3, IL-28B, IL28C

For Research Use Only. Not for use in diagnostic procedures.