missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IFNL3 (IL28B) Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human IFNL3 (NP_742151.2) (Met 1-Val 196) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Invitrogen LS11890H08H50

 View more versions of this product

Catalog No. 11890H08H50



Description

Description

Interferon Lambda 3 (IFNL3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: 1. Measured in anti-viral assays using WISH human amnion cells infected with vesicular stomatitis virus (VSV), the EC50 for this effect is typically 4-20ng/mL; 2. Measured by its binding ability in a functional ELISA; immobilized human IL28B at 20μg/mL (100μL/well) can bind human IL10RB with a linear ranger of 3.125-25μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, 500mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
Interferons|Interleukins
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
21kDa
50μg
<1 EU/μg
Interferon lambda 3
His Tag
Cytokines & Receptors
Assay Standard
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
IFNL3, IL-28B, IL28C
IFNL3
SDS
Documents

For Research Use Only. Not for use in diagnostic procedures.