Please login to your online account to display your discounted pricing

Sino Biological™ IFNGR1 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 245) of human IFN-γ R1 pre-protein (NP_000407.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10338H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10338H08H5

  • / Each
 

Description & Specifications

Specifications

Purification Method PAGE
Purity ≥90% by SDS-PAGE
Gene ID (Entrez) 3459
Structural Form Truncated/Recombinant
Gene Alias IFNGR1, RP3-503F13.3, CD119, FLJ45734, IFNGR
Common Name Interferon gamma receptor 1
Validated Application Assay Standard
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Molecular Weight 27.3kDa
Quantity 5μg
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene Symbol IFNGR1
Protein Family Cytokines & Receptors/CD Proteins
Endotoxin Concentration <1 EU/μg
Termination C-Terminal
Protein Tag His Tag

Interferon Gamma Receptor 1 (IFNGR1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.