missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IFNG Recombinant Mouse Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mouse IFNG (P01580) (Met 1-Cys 155) fused to the Fc region of human IgG1 at the C-terminus

$194.00 - $194.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IFN-g, Ifg
View More Specs

Includes: Five vials of lyophilized protein

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50709M02H50 View Documents Life Technologies
LS50709M02H50
50μg Each for $194.00
 
Description

Description

Interferon gamma (IFNG) recombinant mouse protein is supplied as a lyophilized powder
  • Activity: Measured in anti-viral assays using L929 cells infected with vesicular stomatitisvirus (VSV); the ED50 for this effect is typically 20–80ng/mL
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% mannitol, 5% trehalose, 0.02% Tween™ 80
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
IFN-g, Ifg
PAGE
Lyophilized
15978

For Research Use Only. Not for use in diagnostic procedures.