missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IFNB1 (INF-β) Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human IFNB1 (NP_002167.1), residues 1 to 187, fused to the Fc region of human IgG1 at the C-terminus

$94.75 - $402.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IFNB1, IFB, IFF, IFNB, mgC96956
Gene ID (Entrez) 3456
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10704H02H25 View Documents Life Technologies
LS10704H02H25
5 x 5μg Each for $235.00
 
10704H02H5 View Documents Life Technologies
LS10704H02H5
5μg Each for $94.75
 
10704H02H50 View Documents Life Technologies
LS10704H02H50
50μg Each for $402.00
 
Description

Description

Interferon, beta 1 (IFNB1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.
  • Activity: Measured in anti-viral assays using WISH human amnion cells infected with vesicular stomatitis virus (VSV), the ED50 for this effect is 0.5-2ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
3456
<1 EU/μg
IFNB1, IFB, IFF, IFNB, mgC96956

For Research Use Only. Not for use in diagnostic procedures.