Please login to your online account to display your discounted pricing

Sino Biological™ IFNAR2 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of the human IFNAR2 isoform a (NP_997468.1) (Met 1-Lys 243) fused to a polyhistidine tag at the C-terminus

$109.00 - $1,320.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IFN-R, IFN-alpha-REC, IFNABR, IFNARB
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10359H08H25

DFS Item
 
life technologies
LS10359H08H25
5 x 5μg Each for $236.00

10359H08H5

DFS Item
 
life technologies
LS10359H08H5
5μg Each for $109.00

10359H08HL

DFS Item
 
life technologies
LS10359H08H250
5 x 50μg Each for $1,320.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IFN-R, IFN-alpha-REC, IFNABR, IFNARB

Interferon (alpha, beta and omega) receptor 2 (IFNAR2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by its ability to inhibit rhesus IFNB-mediated protection of WISH human amnion cells infected with vesicular stomatitis virus (VSV) to viral lysis, the EC50 for this effect is typically 0.08-0.4μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.