missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IFNAR1 Recombinant Human Protein, IgG1-Fc Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human IFNAR1 isoform 1 (P17181-1) extracellular domain (Met 1- Lys 436) fused to the Fc region of human IgG1 at the C-terminus

$476.00 - $476.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias AVP, IFN-alpha-REC, IFNAR, IFNBR, IFRC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
13222H02H50 View Documents Life Technologies
LS13222H02H50
50μg Each for $476.00
 
Description

Description

Interferon (alpha, beta and omega) Receptor 1 (IFNAR1), recombinant human protein is supplied as a lyophilized powder.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
AVP, IFN-alpha-REC, IFNAR, IFNBR, IFRC
PAGE
Lyophilized
3454

For Research Use Only. Not for use in diagnostic procedures.