Please login to your online account to display your discounted pricing

Sino Biological™ IFNAR1 Recombinant Human Protein, IgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human IFNAR1 isoform 1 (P17181-1) extracellular domain (Met 1- Lys 436) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS13222H02H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 13222H02H50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 74kDa
Purification Method PAGE
Purity ≥85% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Termination C-Terminal
Structural Form Truncated/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Alias AVP, IFN-alpha-REC, IFNAR, IFNBR, IFRC
Common Name Interferon (alpha, beta and omega) Receptor 1
Gene Symbol IFNAR1
Protein Tag IgG1 Fc Tag
Validated Application Assay Standard
Protein Family Cytokines & Receptors
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 3454

Interferon (alpha, beta and omega) Receptor 1 (IFNAR1), recombinant human protein is supplied as a lyophilized powder.

  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.