missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ IFNA7 Recombinant Human Protein, N-hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human IFNA7 (P01567) (Cys 24-Asp 189) fused to the Fc region of human IgG1 at the N-terminus

$175.00 - $175.00

Specifications

Purity >85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IFN-alphaJ, IFNA-J
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10341H01H50 Life Technologies
LS10341H01H50
50μg Each for $175.00
Specifications

Description

Interferon Alpha 7 (IFNA7), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

>85% by SDS-PAGE
<1 EU/μg
IFN-alphaJ, IFNA-J
PAGE
Lyophilized
3444