missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ IFNA4 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human IFNA4 (NP_066546.1) (Cys 23-Asp 189) fused to the Fc region of human IgG1 at the N-terminus

Manufacturer:  Life Technologies LS10336H01H250

 View more versions of this product

Catalog No. 10336H01HL


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; IFNA4 Recombinant Human Protein, His Tag; 5 x 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Interferon, alpha 4 (IFNA4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.
  • Activity: Measured in anti-viral assays using WISH human amnion cells infected with vesicular stomatitis virus (VSV), the EC50 for this effect is 0.8-4ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
46kDa
5 x 50μg
Full Length/Recombinant
<1 EU/μg
Interferon, alpha 4
Assay Standard
Mammalian
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interferons
INFA4, mgC142200
IFNA4
Cytokines & Receptors
IgG1 Fc Tag