missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ IFNA10 Recombinant Human Protein, N-hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human IFNA10 (P01566) (Cys 24-Asp 189) fused to the Fc region of human IgG1 at the N-terminus

Manufacturer:  Life Technologies LS10349H01H50

 View more versions of this product

Catalog No. 10349H01H50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; IFNA10 Recombinant Human Protein, N-hIgG1-Fc Tag; 50ug; Source: Human; Purity: greater than 85% by SDS-PAGE

Interferon Alpha 10 (IFNA10), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
3446
PAGE
50μg
N-Terminal
<1 EU/μg
Interferon alpha 10
Assay Standard
Mammalian
Lyophilized
48kDa
>85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
IFN-alphaC, mgC119878, mgC119879
IFNA10
Cytokines & Receptors
IgG1 Fc Tag