missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ IDO1 Recombinant Human Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human IDO1 (P14902-1) (Ala 2-Gly 403) fused to a polyhistidine tag at the N-terminus

$225.00 - $1,190.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Format Frozen
Gene Alias IDO, INDO
Gene ID (Entrez) 3620
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11650H07E25 Life Technologies
LS11650H07E25
5 x 5μg Each for $652.00
11650H07E5 Life Technologies
LS11650H07E5
5μg Each for $225.00
11650H07E50 Life Technologies
LS11650H07E50
50μg Each for $1,190.00
Specifications

Description

Indoleamine 2,3-Dioxygenase 1 (IDO1), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: 50mM Tris, 150mM NaCl, 10% Glycerol, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥85% by SDS-PAGE
Frozen
3620
PAGE
IDO, INDO