missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ IDO1 Recombinant Human Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human IDO1 (P14902-1) (Ala 2-Gly 403) fused to a polyhistidine tag at the N-terminus

Manufacturer:  Life Technologies LS11650H07E5

 View more versions of this product

Catalog No. 11650H07E5


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; IDO1 Recombinant Human Protein, N-His Tag; 5ug; Source: Human; Purity: greater than or equal to 85% by SDS-PAGE

Indoleamine 2,3-Dioxygenase 1 (IDO1), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: 50mM Tris, 150mM NaCl, 10% Glycerol, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
3620
PAGE
5μg
Full Length/Recombinant
Indoleamine 2,3-Dioxygenase 1
Assay Standard
E. coli
Frozen
46kDa
≥85% by SDS-PAGE
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
IDO, INDO
IDO1
Other Proteins
His Tag