Please login to your online account to display your discounted pricing

Sino Biological™ IDO1 Recombinant Human Protein, N-His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human IDO1 (P14902-1) (Ala 2-Gly 403) fused to a polyhistidine tag at the N-terminus

Manufacturer: life technologies  LS11650H07E25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11650H07E25

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 3620
Purification Method PAGE
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene Symbol IDO1
Expression System E. coli
Format Frozen
Protein Family Other Proteins
Gene Alias IDO, INDO
Protein Tag His Tag
Storage Requirements Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Purity ≥85% by SDS-PAGE
Common Name Indoleamine 2,3-Dioxygenase 1
Quantity 5 x 5μg
Structural Form Full Length/Recombinant
Validated Application Assay Standard
Molecular Weight 46kDa

Indoleamine 2,3-Dioxygenase 1 (IDO1), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: 50mM Tris, 150mM NaCl, 10% Glycerol, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.