Please login to your online account to display your discounted pricing

Sino Biological™ ICAM1 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human ICAM1 (NP_000192.2) (Met 1 - Glu 480) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10346H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10346H08H50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 51kDa
Purity ≥90% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Truncated/Recombinant/Cell Surface Protein
Protein Subtype Intercellular Adhesion Molecule
Endotoxin Concentration <1 EU/μg
Gene Alias BB2, CD54, P3.58
Common Name Intercellular adhesion molecule 1
Gene Symbol ICAM1
Protein Tag His Tag
Validated Application Assay Standard
Protein Family CD Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Intercellular adhesion molecule 1 (ICAM1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by the ability of the immobilized protein to support the adhesion of PMA stimulated HSB2 human peripheral blood acute lymphoblastic leukemia cells; when cells are added to ICAM1 coated plates (12.5μg/mL, 100μL/well), approximately 20-40% will adhere specifically
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.