Please login to your online account to display your discounted pricing

Sino Biological™ HTRA2 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human HTRA2 (O43464-1) (Ala 134-Glu 458) fused to a polyhistide tag at the C-terminus

$205.00 - $205.00

Specifications

Purity ≥85% by SDS-PAGE
Format Frozen
Gene Alias OMI, PARK13, PRSS25
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10619H08E25

DFS Item
 
life technologies
LS10619H08E25
5 x 5μg Each for $205.00
Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE
Format Frozen
Gene Alias OMI, PARK13, PRSS25

HTRA Serine Peptidase 2 (HTRA2), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Protease activity demonstrated by HTRA2 cleavage of bovine beta-casein (Sigma Cat. No. C-6905); incubation of beta-casein at 0.2μg/mL with Recombinant Human HTRA2 at 0.02μg/mL (ratio of 10:1) for 60 minutes at 45°C in 50mM Tris, pH 8.0, which results in >95% cleavage of beta-casein, as revealed by SDS-PAGE
  • Formulation: 50mM Tris, 0.3M NaCl, 1mM DTT, 0.2μg/μL BSA, 20% glycerol, pH 7.8
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.