Please login to your online account to display your discounted pricing

Sino Biological™ HPX Recombinant Mouse Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mouse HPX (NP_059067.2) (Met 1 - Gln 460) fused to a polyhistidine tag at the C-terminus

$110.00 - $1,124.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Hpxn, hx, HPX
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50271M08H25

DFS Item
 
life technologies
LS50271M08H25
5 x 5μg Each for $241.00

50271M08H5

DFS Item
 
life technologies
LS50271M08H5
5μg Each for $110.00

50271M08H50

DFS Item
 
life technologies
LS50271M08H50
50μg Each for $360.00

50271M08HL

DFS Item
 
life technologies
LS50271M08H250
5 x 50μg Each for $1,124.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Hpxn, hx, HPX

Hemopexin (HPX), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to bind protoporphyrin IX (PPPIX); Recombinant mouse Hemopexin (HPX) binds >15μM PPPIX, resulting in a 50% decrease in the fluorescence signal of mouse HPX
  • Formulation: Lyophilized in 20mM MES, 150mM NaCl, pH 6.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.