missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HPX Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human HPX (NP_000604.1) (Met 1-His 462) fused to a polyhistidine tag at the C-terminus

$222.00 - $222.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias FLJ56652, HX, HPX
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10870H08H25 View Documents Life Technologies
LS10870H08H25
5 x 5μg Each for $222.00
 
Description

Description

Hemopexin (HPX), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind protoporphyrin IX (PPPIX); recombinant human Hemopexin (HPX) binds >25μM PPPIX, resulting in a 50% decrease in the fluorescence signal of human HPX
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
FLJ56652, HX, HPX

For Research Use Only. Not for use in diagnostic procedures.