missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HPGD Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human HPGD (NP_000851.2) (Met 1-Gln 266) fused to a polyhistidine tag at the C-terminus

$235.00 - $235.00

Specifications

Purity ≥90% by SDS-PAGE
Format Frozen
Gene Alias 15-PGDH, PGDH, PGDH1, SDR36C1, HPGD
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11205H08E5 View Documents Life Technologies
LS11205H08E5
5μg Each for $235.00
 
Description

Description

Hydroxyprostaglandin Dehydrogenase 15-(Nad) (HPGD), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measure by the production of NADH during the oxidation of PGF2-alpha; the specific activity is >1500pmole/min./μg
  • Formulation: 50mM Tris, 100mM NaCl, 0.5mM DTT, 0.2μg/μL BSA, 10% glycerol, pH.7.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Frozen
15-PGDH, PGDH, PGDH1, SDR36C1, HPGD

For Research Use Only. Not for use in diagnostic procedures.