Please login to your online account to display your discounted pricing

Sino Biological™ HPGD Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human HPGD (NP_000851.2) (Met 1-Gln 266) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11205H08E5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11205H08E5

  • / Each
 

Description & Specifications

Specifications

Gene Symbol HPGD
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Molecular Weight 29.7kDa
Purity ≥90% by SDS-PAGE
Quantity 5μg
Storage Requirements Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Other Proteins
Gene Alias 15-PGDH, PGDH, PGDH1, SDR36C1, HPGD
Common Name Hydroxyprostaglandin dehydrogenase 15-(NAD)
Protein Tag His Tag
Validated Application Assay Standard
Expression System E. coli
Format Frozen

Hydroxyprostaglandin Dehydrogenase 15-(Nad) (HPGD), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measure by the production of NADH during the oxidation of PGF2-alpha; the specific activity is >1500pmole/min./μg
  • Formulation: 50mM Tris, 100mM NaCl, 0.5mM DTT, 0.2μg/μL BSA, 10% glycerol, pH.7.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.