missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ HIST2H2BE Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human HIST2H2BE (Q16778) (Met 1-Lys 126)

$584.00 - $930.00

Specifications

Purity ≥95% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias GL105, H2B, H2B.1, H2BFQ, H2BGL105, H2BQ, mgC119802, mgC119804, mgC129733
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11362HNAE25 Life Technologies
LS11362HNAE25
5 x 5μg Each for $584.00
11362HNAE50 Life Technologies
LS11362HNAE50
50μg Each for $930.00
Specifications

Description

Histone Cluster 2, H2BE (HIST2H2BE), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 2mM β-Mercaptoethanol, pH 6.9, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥95% by SDS-PAGE
<1 EU/μg
GL105, H2B, H2B.1, H2BFQ, H2BGL105, H2BQ, mgC119802, mgC119804, mgC129733
PAGE
Lyophilized
8349