Please login to your online account to display your discounted pricing

Sino Biological™ HIST2H2BE Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human HIST2H2BE (Q16778) (Met 1-Lys 126)

Manufacturer: life technologies  LS11362HNAE50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11362HNAE50

  • / Each
 

Description & Specifications

Specifications

Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Molecular Weight 14.2kDa
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Endotoxin Concentration <1 EU/μg
Validated Application Assay Standard
Gene Symbol HIST2H2BE
Gene ID (Entrez) 8349
Purification Method PAGE
Purity ≥95% by SDS-PAGE
Gene Alias GL105, H2B, H2B.1, H2BFQ, H2BGL105, H2BQ, mgC119802, mgC119804, mgC129733
Common Name Histone Cluster 2, H2BE
Protein Family Histones
Structural Form Full Length/Recombinant
Format Lyophilized

Histone Cluster 2, H2BE (HIST2H2BE), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 2mM β-Mercaptoethanol, pH 6.9, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.