Please login to your online account to display your discounted pricing

Sino Biological™ HIST1H3A Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the native human HIST1H3A (P6843) (Met 1-Ala 136)

Manufacturer: life technologies  LS11231HNAE50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11231HNAE50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 15.5kDa
Purification Method PAGE
Purity ≥95% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Structural Form Full Length/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Alias HIST1H3A, H3/A, H3FA, HIST1H3B, HIST1H3C, HIST1H3D, HIST1H3E, HIST1H3F, HIST1H3G
Common Name Histone cluster 1, H3a
Gene Symbol HIST1H3A
Validated Application Assay Standard
Protein Family Histones
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 8350

Histone H3.1 (HIST1H3A), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 2mM β-Mercaptoethanol, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.