Please login to your online account to display your discounted pricing

Sino Biological™ HA1 Subunit Recombinant Influenza A Virus Protein, Subtype H5N1, (A/Vietnam/1203/2004), mIgG1-Fc.His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the N-terminal segment (Met 1-Arg 342) of influenza A virus, subtype H5N1 (A/VietNam/1203/2004), HA1 subunit (AAW80717.1) fused to the His-tagged Fc region of mouse IgG1 at the C-terminus

$120.00 - $1,076.00

Specifications

Purity ≥85% by SDS-PAGE and SEC-HPLC Analysis
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HA1
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10003V06H1

DFS Item
 
life technologies
LS10003V06H150
50μg Each for $353.00

10003V06H15

DFS Item
 
life technologies
LS10003V06H15
5μg Each for $120.00

10003V06H1L

DFS Item
 
life technologies
LS10003V06H1250
5 x 50μg Each for $1,076.00
Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE and SEC-HPLC Analysis
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HA1

H5N1, recombinant influenza A virus protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.

  • Activity: Measured by its binding ability in a functional ELISA
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.