Please login to your online account to display your discounted pricing

Sino Biological™ HA1 Subunit Recombinant Influenza A Virus Protein, Subtype H5N1 (A/Anhui/1/2005), His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the N-terminal segment (Met 1-Pro 337) of influenza A virus, subtype H5N1 (A/Anhui/1/2005), HA1 subunit (ABD28180.1) fused to a polyhistidine tag at the C-terminus

$120.00 - $1,078.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HA1
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11048V08H25

DFS Item
 
life technologies
LS11048V08H25
5μg Each for $120.00

11048V08H22

DFS Item
 
life technologies
LS11048V08H225
5 x 5μg Each for $227.00

11048V08H2L

DFS Item
 
life technologies
LS11048V08H2250
5 x 50μg Each for $1,078.00

11048V08HL

DFS Item
 
life technologies
LS11048V08H250
50μg Each for $353.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HA1

Hemagglutinin, Influenza A Virus (HA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.