Please login to your online account to display your discounted pricing

Sino Biological™ HA1 Subunit Recombinant Influenza A Virus Protein, Subtype H3N2 (A/Aichi/2/1968), His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the N-terminal segment (Met 1-Arg 345) of the influenza A virus, subtype H3N2 (A/Aichi/2/1968), HA1 (AAA43178.1) fused to a C-terminal polyhistidine tag

Manufacturer: life technologies  LS11707V08H15

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11707V08H15

  • / Each
 

Description & Specifications

Specifications

Format Lyophilized
Expression System Mammalian
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Molecular Weight 37.6kDa
Quantity 5μg
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Protein Tag His Tag
Protein Subtype Viral Proteins
Structural Form Truncated/Recombinant/Cell Surface Protein
Protein Family Other Proteins
Gene Symbol HA
Gene Alias HA1
Common Name Hemagglutinin
Validated Application Assay Standard
Endotoxin Concentration <1 EU/μg
Purity ≥90% by SDS-PAGE

Hemagglutinin, Influenza A Virus, HA1 Subunit (HA1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.