Please login to your online account to display your discounted pricing

Sino Biological™ HA1 Subunit Recombinant Influenza A Virus Protein, Subtype H1N1 (A/California/04/2009), His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding an N-terminal segment (Met 1-Arg 344) of influenza A virus, subtype H1N1 (A/California/04/2009), HA1 (ACP41105.1) fused to a C-terminal polyhistidine tag

$120.00 - $1,052.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HA1
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11055V08H42

DFS Item
 
life technologies
LS11055V08H425
5 x 5μg Each for $226.00

11055V08H45

DFS Item
 
life technologies
LS11055V08H45
5μg Each for $120.00

11055V08H4L

DFS Item
 
life technologies
LS11055V08H4250
5 x 50μg Each for $1,052.00

11055V08H4S

DFS Item
 
life technologies
LS11055V08H450
50μg Each for $351.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HA1

Hemagglutinin, Influenza A Virus (HA1 Subunit), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.