Please login to your online account to display your discounted pricing

Sino Biological™ HA1 Subunit Recombinant Influenza A Virus Protein, Subtype H1N1 (A/California/04/2009), His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding an N-terminal segment (Met 1-Arg 344) of influenza A virus, subtype H1N1 (A/California/04/2009), HA1 (ACP41105.1) fused to a C-terminal polyhistidine tag

Manufacturer: life technologies  LS11055V08H450

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11055V08H4S

  • / Each
 

Description & Specifications

Specifications

Endotoxin Concentration <1 EU/μg
Expression System Mammalian
Format Lyophilized
Structural Form Truncated/Recombinant/Cell Surface Protein
Protein Family Other Proteins
Common Name Hemagglutinin
Quantity 50μg
Gene Alias HA1
Protein Tag His Tag
Protein Subtype Viral Proteins
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Validated Application Assay Standard
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Molecular Weight 38kDa
Purity ≥90% by SDS-PAGE
Gene Symbol HA

Hemagglutinin, Influenza A Virus (HA1 Subunit), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.