missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HA Recombinant Influenza A Virus Protein, Subtype H9N2 (A/Hong Kong/1073/99), His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Lys 523) of influenza A virus, sybtype H9N2 (A/Hong Kong/1073/99 (H9N2)), HA (NP_859037.1) fused to a polyhistidine tag at the C-terminus

$1,110.00 - $1,110.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HA
Gene ID (Entrez) 1460996
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11229V08HL View Documents Life Technologies
LS11229V08H250
5 x 50μg Each for $1,110.00
 
Description

Description

Hemagglutinin, Influenza A Virus (HA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Subunits HA1 and HA2 remain uncleaved
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
1460996
<1 EU/μg
HA

For Research Use Only. Not for use in diagnostic procedures.