missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ HA Recombinant Influenza A Virus Protein, Subtype H9N2 (A/Hong Kong/1073/99), His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Lys 523) of influenza A virus, sybtype H9N2 (A/Hong Kong/1073/99 (H9N2)), HA (NP_859037.1) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS11229V08H250

 View more versions of this product

Catalog No. 11229V08HL


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; HA Recombinant Influenza A Virus Protein, subtype H9N2 (A/Hong Kong/1073/99), His Tag; 5 x 50ug; Source: Influenza; Purity: greater than or equal to 90% by SDS-PAGE

Hemagglutinin, Influenza A Virus (HA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Subunits HA1 and HA2 remain uncleaved
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
1460996
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Viral Proteins
HA
HA
Other Proteins
His Tag
Lyophilized
58kDa
5 x 50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Hemagglutinin
Assay Standard
Mammalian