Please login to your online account to display your discounted pricing

Sino Biological™ HA Recombinant Influenza A Virus Protein, Subtype H7N7 (A/Netherlands/219/03), His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the native extracellular domain (Met 8-Phe 544) of influenza A virus, subtype H7N7 (A/Netherlands/219/03), HA (AAR02640.1) (Met 8-Phe 544), fused to a polyhistidine tag at the C-terminus

$227.00 - $353.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HA
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11082V08B25

DFS Item
 
life technologies
LS11082V08B25
5 x 5μg Each for $227.00

11082V08B50

DFS Item
 
life technologies
LS11082V08B50
50μg Each for $353.00
Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HA

Hemagglutinin, Influenza A Virus (HA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured in hemagglutination test using 1% chicken erythrocyte; the HA titer is 0.1-0.5μg/mL for 1% chicken red blood cells
  • Formulation: Lyophilized in 20mM Tris, pH 7.4, 500mM NaCl, 10% glycerol, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.