Please login to your online account to display your discounted pricing

Sino Biological™ HA Recombinant Influenza A Virus Protein, Subtype H7N7 (A/Netherlands/219/03), His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the native extracellular domain (Met 8-Phe 544) of influenza A virus, subtype H7N7 (A/Netherlands/219/03), HA (AAR02640.1) (Met 8-Phe 544), fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11082V08B25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11082V08B25

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Format Lyophilized
Common Name Hemagglutinin
Molecular Weight 59kDa
Quantity 5 x 5μg
Endotoxin Concentration <1 EU/μg
Protein Subtype Viral Proteins
Protein Family Other Proteins
Structural Form Truncated/Recombinant/Cell Surface Protein
Gene Alias HA
Validated Application Assay Standard
Gene Symbol HA
Purity ≥85% by SDS-PAGE
Expression System Insect
Protein Tag His Tag
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

Hemagglutinin, Influenza A Virus (HA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured in hemagglutination test using 1% chicken erythrocyte; the HA titer is 0.1-0.5μg/mL for 1% chicken red blood cells
  • Formulation: Lyophilized in 20mM Tris, pH 7.4, 500mM NaCl, 10% glycerol, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.