Please login to your online account to display your discounted pricing

Sino Biological™ HA Recombinant Influenza A Virus Protein, Subtype H5N1 (A/Indonesia/5/2005), His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 531) of influenza A virus, subtype H5N1 (A/Indonesia/5/2005), HA (ABW06108.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11060V08H15

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11060V08H15

  • / Each
 

Description & Specifications

Specifications

Format Lyophilized
Quantity 5μg
Molecular Weight 59kDa
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Gene Alias HA
Gene Symbol HA
Protein Tag His Tag
Protein Family Other Proteins
Structural Form Truncated/Recombinant/Cell Surface Protein
Protein Subtype Viral Proteins
Validated Application Assay Standard
Common Name Hemagglutinin
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Expression System Mammalian

Hemagglutinin, Influenza A Virus (HA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • The protein has its cleavage site mutated (RERRRKKR to TETR), therefore, the HA1 and HA2 subunits remain uncleaved
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.