missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HA Recombinant Influenza A Virus Protein, Subtype H5N1 (A/Egypt/N05056/2009), His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 530) of the influenza A virus, subtype H5N1 (A/Egypt/N05056/2009), HA (ACT15357.1) fused to a polyhistidine tag at the C-terminus

$1,120.00 - $1,120.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11702V08HL View Documents Life Technologies
LS11702V08H250
5 x 50μg Each for $1,120.00
 
Description

Description

Hemagglutinin, Influenza A Virus (HA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • The protein has its cleavage site mutated (RERRRKKR to TETR), therefore, the HA1 and HA2 subunits remain uncleaved
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in 20mM Tris, pH 7.4, 500mM NaCl, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
HA

For Research Use Only. Not for use in diagnostic procedures.