Please login to your online account to display your discounted pricing

Sino Biological™ HA Recombinant Influenza A Virus Protein, Subtype H5N1 (A/Anhui/1/2005), His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 527) of influenza A virus, subtype H5N1 (A/Anhui/1/2005), HA (ABD28180.1) fused to a polyhistidine tag at the C-terminus

$120.00 - $1,078.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HA
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11048V08H15

DFS Item
 
life technologies
LS11048V08H15
5μg Each for $120.00

11048V08H

DFS Item
 
life technologies
LS11048V08H150
50μg Each for $353.00

11048V08H1L

DFS Item
 
life technologies
LS11048V08H1250
5 x 50μg Each for $1,078.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HA

Hemagglutinin, Influenza A Virus (HA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • The protein has its cleavage site mutated (RERRRKKR to TETR), therefore, the HA1 and HA2 subunits remain uncleaved
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.