Please login to your online account to display your discounted pricing

Sino Biological™ HA Recombinant Influenza A Virus Protein Subtype H4N8 (A/Chicken/Alabama/1/1975), His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of influenza A virus hemagglutinin (A/chicken/Alabama/1/1975(H4N8) P19695 (Met 1-Ile528) fused to a polyhistidine tag at the C-terminus

$365.00 - $365.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HA
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

40025V08H50

DFS Item
 
life technologies
LS40025V08H50
50μg Each for $365.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HA
Gene ID (Entrez) Unknown

Hemagglutinin A (HA), recombinant H4N8 (A/chicken/Alabama/1/1975) HA protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.