Please login to your online account to display your discounted pricing

Sino Biological™ HA Recombinant Influenza A Virus Protein, subtype H1N1, (A/Brisbane/59/2007), His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 528) of influenza A virus, subtype H1N1 (A/Brisbane/59/2007), HA (ACA28844.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11052V08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11052V08H5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 59kDa
Purity ≥90% by SDS-PAGE
Quantity 5μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Truncated/Recombinant/Cell Surface Protein
Protein Subtype Viral Proteins
Endotoxin Concentration <1 EU/μg
Gene Alias HA
Common Name Hemagglutinin
Gene Symbol HA
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Other Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Hemagglutinin, Influenza A Virus (HA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Subunits HA1 and HA2 remain uncleaved
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.