Please login to your online account to display your discounted pricing

Sino Biological™ HA Recombinant Influenza A, Subtype H3N2 (A/Perth/16/2009) Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of influenza A virus hemagglutinin (A/Perth/16/2009(H3N2)) (Met 1-Trp 530) (ACS71642.1) fused to a polyhistidine tag at the C-terminus

$1,134.00 - $1,134.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HA
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

40043V08HL

DFS Item
 
life technologies
LS40043V08H250
5 x 50μg Each for $1,134.00
Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias HA

Hemagglutinin (HA), recombinant viral protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.