Please login to your online account to display your discounted pricing

Sino Biological™ HA Recombinant Influenza A, Subtype H2N2 (A/Canada/720/2005) Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 516) of the influenza hemagglutinin (A/Canada/720/2005 (H2N2)) (AAY28987.1) (HA1+HA2, uncleaved) fused to a C-terminal polyhistidine tag

Manufacturer: life technologies  LS11688V08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11688V08H25

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene Symbol HA
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Format Lyophilized
Structural Form Truncated/Recombinant
Expression System Mammalian
Protein Tag His Tag
Termination C-Terminal
Common Name Hemagglutinin
Validated Application Assay Standard
Molecular Weight 59kDa
Endotoxin Concentration <1 EU/μg
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Alias HA
Protein Family Cell Structure/Organelle Proteins

Hemagglutinin (HA), recombinant viral protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.