missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ HA, 203 Asp/Asn Recombinant Influenza A Virus Protein, Subtype H1N1 (A/New Caledonia/20/1999), His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 528) of the influenza A virus, subtype H1N1 (A/New Caledonia/20/1999), HA (CAC86622.1), containing the 203 Asp/Asn mutation, fused to a C-terminal polyhistidine tag

Manufacturer:  Life Technologies LS11683V08H5

 View more versions of this product

Catalog No. 11683V08H5


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; HA, 203 Asp/Asn Recombinant Influenza A Virus Protein, subtype H1N1 (A/New Caledonia/20/1999), His tag; 5ug; Source: Influenza; Purity: greater than or equal to 95% by SDS-PAGE

Hemagglutinin, Influenza A Virus (HA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Subunits HA1 and HA2 remain uncleaved
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
59kDa
5μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Hemagglutinin
Assay Standard
Mammalian
Lyophilized
≥95% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Viral Proteins
HA
HA
Other Proteins
His Tag