Please login to your online account to display your discounted pricing

Sino Biological™ HA, 203 Asp/Asn Recombinant Influenza A Virus Protein, Subtype H1N1 (A/New Caledonia/20/1999), His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 528) of the influenza A virus, subtype H1N1 (A/New Caledonia/20/1999), HA (CAC86622.1), containing the 203 Asp/Asn mutation, fused to a C-terminal polyhistidine tag

Manufacturer: life technologies  LS11683V08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11683V08H25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 59kDa
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Purity ≥95% by SDS-PAGE
Quantity 5 x 5μg
Structural Form Truncated/Recombinant/Cell Surface Protein
Protein Subtype Viral Proteins
Endotoxin Concentration <1 EU/μg
Gene Alias HA
Common Name Hemagglutinin
Gene Symbol HA
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Other Proteins
Format Lyophilized

Hemagglutinin, Influenza A Virus (HA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Subunits HA1 and HA2 remain uncleaved
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.