missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GRN Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human GRN (NP_002078.1) (Met 1-Leu 593) fused to a C-terminal polyhistidine tag was expressed

$99.50 - $99.50

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias GRN, GEP, GP88, PCDGF, PEPI, PGRN
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10826H08H5 View Documents Life Technologies
LS10826H08H5
5μg Each for $99.50
 
Description

Description

Granulin (GRN), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
GRN, GEP, GP88, PCDGF, PEPI, PGRN
PAGE
Lyophilized
2896

For Research Use Only. Not for use in diagnostic procedures.