missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GPC5 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human GPC5 (NP_004457.1) (Met 1-Thr 554) fused to the polyhistidine tag at the C-terminus

$399.00 - $399.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias GPC5, bA93M14.1, glypican proteoglycan 5, glypican-5
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10079H08B50 View Documents Life Technologies
LS10079H08B50
50μg Each for $399.00
 
Description

Description

Glypican 5 (GPC5), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human GPC5 at 5μg/mL (100μL/well) can bind human FGF2 with a linear range of 0.156-2.5ng/mL
  • Formulation: Lyophilized in 50mM Tris, 100mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Frozen
GPC5, bA93M14.1, glypican proteoglycan 5, glypican-5

For Research Use Only. Not for use in diagnostic procedures.