missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ GPC5 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human GPC5 (NP_004457.1) (Met 1-Thr 554) fused to the polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS10079H08B5

 View more versions of this product

Catalog No. 10079H08B5


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; GPC5 Recombinant Human Protein, His Tag; 5ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Glypican 5 (GPC5), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human GPC5 at 5μg/mL (100μL/well) can bind human FGF2 with a linear range of 0.156-2.5ng/mL
  • Formulation: Lyophilized in 50mM Tris, 100mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
60.5kDa
5μg
Proteoglycans
GPC5, bA93M14.1, glypican proteoglycan 5, glypican-5
GPC5
Other Proteins
His Tag
Frozen
≥90% by SDS-PAGE
Store at -80 °C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Glypican 5
Assay Standard
Insect