Please login to your online account to display your discounted pricing

Sino Biological™ GPC3 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human GPC3 (P51654-1) (Met 1-His 559) fused to a C-terminal polyhistidine tag

Manufacturer: life technologies  LS10088H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10088H08H5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 62kDa
Purity ≥85% by SDS-PAGE
Quantity 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Proteoglycans
Endotoxin Concentration <1 EU/μg
Gene Alias GPC3, AC002420.1, DGSX, GTR2-2, MXR7, OCI-5, SDYS, SGB, SGBS, SGBS1
Common Name Glypican 3
Gene Symbol GPC3
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Cell Structure/Organelle Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Glypican 3 (GPC3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human GPC3 at 5μg/mL (100μL/well) can bind human bFGF/FGF2 with a linear range of 0.156-2.5ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.